(S)-4-Boc-(3-hydroxymethyl)morpholine 97%
|
Warning
|
Synonyms:
tert-Butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate
H64574-MD
H64574-03
H64574-06
AAH64574-MDEA
66.44
USD
AAH64574-MD
AAH64574-06
AAH64574-03
(S)-4-Boc-(3-hydroxymethyl)morpholine 97%
(S)-4-Boc-(3-hydroxymethyl)morpholine
Formula:
C10H19NO4 MW: 217.27 g/mol Melting Pt: 80 °C (lit.) Storage Temperature: Refrigerator |
MDL Number:
MFCD06799478 CAS Number: 714971-28-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...