5-Bromo-3-chloro-2-fluoropyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

H64547-06 H64547-MD H64547-03
AAH64547-06EA 301.99 USD
AAH64547-06 AAH64547-MD AAH64547-03
5-Bromo-3-chloro-2-fluoropyridine 95%
5-Bromo-3-chloro-2-fluoropyridine
Formula: C5H2BrClFN
MW: 210.43 g/mol
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD09834365
CAS Number: 38185-56-7

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR