3-(4-Bromophenyl)-9-phenylcarbazole 98%
|
Warning
|
Synonyms:
3-(4-Bromophenyl)-9-phenyl-9H-carbazole
H64542-MD
H64542-03
H64542-06
AAH64542-06EA
237.54
USD
AAH64542-06
AAH64542-MD
AAH64542-03
3-(4-Bromophenyl)-9-phenylcarbazole 98%
3-(4-Bromophenyl)-9-phenylcarbazole
Formula:
C₂₄H₁₆BrN MW: 398.30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14582939 CAS Number: 1028647-93-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...