2,3-Difluoro-6-methylpyridine 98%

Warning

Synonyms: 2,3-Difluoro-6-picoline

H64514-06 H64514-03 H64514-MD
AAH64514-06EA 270.89 USD
AAH64514-06 AAH64514-MD AAH64514-03
2,3-Difluoro-6-methylpyridine 98%
2,3-Difluoro-6-methylpyridine
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16610828
CAS Number: 1227579-04-5

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR