2-Chloro-3-fluoro-6-methylpyridine 95%
|
Warning
|
H64473-03
H64473-MD
AAH64473-03EA
307.47
USD
AAH64473-03
AAH64473-MD
2-Chloro-3-fluoro-6-methylpyridine 95%
2-Chloro-3-fluoro-6-methylpyridine
Formula:
C₆H₅ClFN MW: 145.56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03095216 CAS Number: 374633-32-6 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...