5-Fluoro-3-nitropyridin-2(1H)-one 98%

Warning

Synonyms: 5-Fluoro-2-hydroxy-3-nitropyridine

H64441-MD H64441-06 H64441-03
AAH64441-MDEA 26.38 USD
AAH64441-MD AAH64441-06 AAH64441-03
5-Fluoro-3-nitropyridin-2(1H)-one 98%
5-Fluoro-3-nitropyridin-2(1H)-one
Formula: C5H3FN2O3
MW: 158.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662412
CAS Number: 136888-20-5

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR