Methyl-6-bromo-5-fluoronicotinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 6-bromo-5-fluoronicotinate

H64395-MD H64395-06 H64395-03
AAH64395-MDEA 32.38 USD
AAH64395-MD AAH64395-06 AAH64395-03
Methyl-6-bromo-5-fluoronicotinate 98%
Methyl-6-bromo-5-fluoronicotinate
Formula: C₇H₅BrFNO₂
MW: 234.024 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698110
CAS Number: 1214336-88-5

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR