Methyl-5-bromosalicylate 98+%
|
Warning
|
Synonyms:
Methyl 5-bromo-2-hydroxybenzoate
H64372-06
H64372-14
H64372-06
AAH64372-06EA
68.5
USD
AAH64372-06
AAH64372-14
CAAAH64372-06
Methyl-5-bromosalicylate 98+%
Methyl 5-bromosalicylate
Formula:
C₈H₇BrO₃ MW: 231.05 g/mol Melting Pt: 61…65 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00016460 CAS Number: 4068-76-2 EINECS: 223-776-4 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...