2-(3-Bromophenyl)naphthalene 98%

H64322-MD H64322-06 H64322-03
AAH64322-03EA 66.44 USD
AAH64322-03 AAH64322-MD AAH64322-06
2-(3-Bromophenyl)naphthalene 98%
2-(3-Bromophenyl)naphthalene
Formula: C₁₆H₁₁Br
Storage Temperature: Ambient
MDL Number: MFCD16658911
CAS Number: 667940-23-0

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR