4-Methoxy-2-methylbenzaldehyde 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64295-03
H64295-06
AAH64295-03EA
45.19
USD
AAH64295-03
AAH64295-06
4-Methoxy-2-methylbenzaldehyde 95%
4-Methoxy-2-methylbenzaldehyde
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 262…267° lit. Density: 1.107 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02261771 CAS Number: 52289-54-0 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...