4-Chloro-2,6-dimethyl-3-nitropyridine 98%

Warning

H64180-06 H64180-MD H64180-03
AAH64180-06EA 516.15 USD
AAH64180-06 AAH64180-MD AAH64180-03
4-Chloro-2,6-dimethyl-3-nitropyridine 98%
4-Chloro-2,6-dimethyl-3-nitropyridine
Formula: C₇H₇ClN₂O₂
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD08543468
CAS Number: 15513-48-1

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR