2-Fluoro-3-methyl-5-pyridineboronic acid 97%

Warning

Synonyms: 2-Fluoro-3-methylpyridine-5-boronic acid(6-Fluoro-5-methylpyridin-3-yl)boronic acid6-Fluoro-5-methyl-3-pyridylboronic acid

H64157-03 H64157-MD H64157-06
AAH64157-03EA 91.77 USD
AAH64157-03 AAH64157-MD AAH64157-06
2-Fluoro-3-methyl-5-pyridineboronic acid 97%
2-Fluoro-3-methyl-5-pyridineboronic acid
Formula: C₆H₇BFNO₂
MW: 154.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411562
CAS Number: 904326-92-7

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR