Morpholin-2-ylmethanol hydrochloride 95%

Warning

Synonyms: 2-Hydroxymethylmorpholine hydrochloride

H64154-03 H64154-MD
AAH64154-MDEA 180.93 USD
AAH64154-MD AAH64154-03
Morpholin-2-ylmethanol hydrochloride 95%
Morpholin-2-ylmethanol hydrochloride
Formula: C5H12ClNO2
MW: 153.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13191721
CAS Number: 144053-98-5

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR