4-Bromo-2-(methylthio)pyrimidine 97%

Danger

Synonyms: 4-Bromo-2-methylsulfanyl-pyrimidine4-Bromo-2-methylsulphanyl-pyrimidine

H64124-MD H64124-06 H64124-03
AAH64124-03EA 109.44 USD
AAH64124-03 AAH64124-06 AAH64124-MD
4-Bromo-2-(methylthio)pyrimidine 97%
4-Bromo-2-(methylthio)pyrimidine
Formula: C5H5BrN2S
MW: 205.07 g/mol
Melting Pt: 30…40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09878932
CAS Number: 959236-97-6
UN: 1759
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR