1,3-Dibromo-5-(trifluoromethoxy)benzene 97%
|
Warning
|
H64115-06
H64115-22
H64115-14
AAH64115-14EA
66.44
USD
AAH64115-14
AAH64115-06
AAH64115-22
1,3-Dibromo-5-(trifluoromethoxy)benzene 97%
1,3-Dibromo-5-(trifluoromethoxy)benzene
Formula:
C₇H₃Br₂F₃O MW: 319.90 g/mol Boiling Pt: 83…85 °C/20 mm lit. Flash Pt: >113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075280 CAS Number: 207226-31-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...