4-Fluoro-3-methylbenzenesulfonyl chloride 97%
|
Danger
|
H64110-MD
H64110-06
H64110-03
AAH64110-03EA
572.85
USD
AAH64110-03
AAH64110-06
AAH64110-MD
4-Fluoro-3-methylbenzenesulfonyl chloride 97%
4-Fluoro-3-methylbenzenesulphonyl chloride
Formula:
C₇H₆ClFO₂S Melting Pt: 35…40 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01365818 CAS Number: 629672-19-1 UN: 3261 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...