2-Bromo-4'-(trifluoromethoxy)acetophenone 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Trifluoromethoxy)phenacyl bromide2-Bromo-1-(4-(trifluoromethoxy)phenyl)ethanone

H64105-06 H64105-03 H64105-MD
AAH64105-MDEA 25.69 USD
AAH64105-MD AAH64105-03 AAH64105-06
2-Bromo-4'-(trifluoromethoxy)acetophenone 97%
2-Bromo-4'-(trifluoromethoxy)acetophenone
Formula: C₉H₆BrF₃O₂
MW: 283.04 g/mol
Melting Pt: 48…55 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00052333
CAS Number: 103962-10-3
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR