Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-((dimethylamino)methylene)-3-oxobutanoateEthyl 2-acetyl-3-(dimethylamino)acrylate

H64073-06 H64073-14 H64073-03
AAH64073-14EA 504.99 USD
AAH64073-14 AAH64073-03 AAH64073-06
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate 95%
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate
Formula: C9H15NO3
MW: 185.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00087563
CAS Number: 51145-57-4

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR