2-(Piperidin-4yl)acetic acid hydrochloride 95%
|
Warning
|
Synonyms:
4-Piperidineacetic acid hydrochloride
H64031-06
H64031-03
H64031-MD
AAH64031-03EA
66.44
USD
AAH64031-03
AAH64031-06
AAH64031-MD
2-(Piperidin-4yl)acetic acid hydrochloride 95%
2-(Piperidin-4yl)acetic acid hydrochloride
Formula:
C7H14ClNO2 MW: 179.64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03414187 CAS Number: 73415-84-6 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...