9-Bromo-10-(naphthalen-1-yl)anthracene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 9-Bromo-10-(1-naphthyl)anthracene

H64024-06 H64024-MD H64024-03
AAH64024-MDEA 30.89 USD
AAH64024-MD AAH64024-06 AAH64024-03
9-Bromo-10-(naphthalen-1-yl)anthracene 98%
9-Bromo-10-(naphthalen-1-yl)anthracene
Formula: C₂₄H₁₅Br
MW: 383.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11046571
CAS Number: 400607-04-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR