3-(2-Bromophenyl)-1-propanol 98%

Warning

Synonyms: 3-(2-Bromo-phenyl)-propan-1-ol

H63884-03 H63884-MD H63884-06
AAH63884-03EA 145.41 USD
AAH63884-03 AAH63884-06 AAH63884-MD
3-(2-Bromophenyl)-1-propanol 98%
3-(2-Bromophenyl)-1-propanol
Formula: C₉H₁₁BrO
MW: 215.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09028685
CAS Number: 52221-92-8

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR