3-(2-Pyridyl)-L-alanine dihydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-2-Pal-OH·2HCl

H63770-03 H63770-MD H63770-06
AAH63770-MDEA 38.38 USD
AAH63770-MD AAH63770-06 AAH63770-03
3-(2-Pyridyl)-L-alanine dihydrochloride 95%
3-(2-Pyridyl)-L-alanine dihydrochloride
Formula: C₈H₁₂Cl₂N₂O₂
MW: 239.096 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06809729
CAS Number: 1082692-96-3

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR