α,3-Dibromo-5-fluorotoluene 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Bromo-5-fluorobenzyl bromide
, 1-Bromo-3-(bromomethyl)-5-fluorobenzene
H63769-03
H63769-06
AAH63769-03EA
233.34
USD
AAH63769-03
AAH63769-06
α,3-Dibromo-5-fluorotoluene 95%
α,3-Dibromo-5-fluorotoluene
Formula:
C₇H₅Br₂F MW: 267.92 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09258959 CAS Number: 216755-57-6 UN: 3261 ADR: "8",III |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...