1,3-Dimethyl-2-thiohydantoin 95%

Warning

Synonyms: 1,3-Dimethyl-2-thioxoimidazolidin-4-one

H63384-06 H63384-MD H63384-03
AAH63384-06EA 1017.87 USD
AAH63384-06 AAH63384-MD AAH63384-03
1,3-Dimethyl-2-thiohydantoin 95%
1,3-Dimethyl-2-thiohydantoin
Formula: C₅H₈N₂OS
MW: 144.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00186242
CAS Number: 1801-62-3

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR