(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-Cit-OHNalpha-Fmoc-L-citrulline

H63342-14 H63342-03 H63342-06
AAH63342-06EA 86.89 USD
AAH63342-06 AAH63342-14 AAH63342-03
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid
Formula: C21H23N3O5
MW: 397.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151943
CAS Number: 133174-15-9

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR