4-Fluoro-3-nitrophenylboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(4-Fluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H62720-MD H62720-03 H62720-06
AAH62720-03EA 69.69 USD
AAH62720-03 AAH62720-MD AAH62720-06
4-Fluoro-3-nitrophenylboronic acid pinacol ester 96%
4-Fluoro-3-nitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BFNO₄
MW: 267.06 g/mol
Melting Pt: 64…65 °C
Storage Temperature: Ambient
MDL Number: MFCD12546515
CAS Number: 1218791-09-3

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR