α,2-Dibromo-4-fluorotoluene 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Bromo-4-fluorobenzylbromide
, 2-Bromo-4-fluorobenzyl bromide
H62157-03
H62157-06
AAH62157-06EA
218.99
USD
AAH62157-06
AAH62157-03
α,2-Dibromo-4-fluorotoluene 96%
α,2-Dibromo-4-fluorotoluene
Formula:
C₇H₅Br₂F MW: 267.92 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03840524 CAS Number: 61150-57-0 UN: 3261 ADR: "8",III |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...