Methyl-2-methyl-5-nitrobenzoate 98%

Synonyms: Methyl 2-methyl-5-nitrobenzoate

H60938-MD H60938-MD
AAH60938-MDEA 666.67 USD
AAH60938-MD CAAAH60938-MD
Methyl-2-methyl-5-nitrobenzoate 98%
Methyl 2-methyl-5-nitrobenzoate
Formula: C₉H₉NO₄
MW: 195.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00230865
CAS Number: 77324-87-9

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR