N-(4-Fluorophenyl)-3-nitrobenzamide 97%

Warning

H60889-03 H60889-MD H60889-MD H60889-03
AAH60889-03EA 481.74 USD
AAH60889-03 CAAAH60889-MD AAH60889-MD CAAAH60889-03
N-(4-Fluorophenyl)-3-nitrobenzamide 97%
N-(4-Fluorophenyl)-3-nitrobenzamide
Formula: C₁₃H₉FN₂O₃
MW: 260.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00034540
CAS Number: 33489-69-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR