(4-Bromophenoxy)acetic acid 95%
Synonyms:
4-Bromophenoxyacetic acid
H60888-06
H60888-14
H60888-14
H60888-06
AAH60888-06EA
32.44
USD
AAH60888-06
AAH60888-14
CAAAH60888-14
CAAAH60888-06
(4-Bromophenoxy)acetic acid 95%
(4-Bromophenoxy)acetic acid
Formula:
(C₆H₄Br)OCH₂COOH MW: 231.05 g/mol Melting Pt: 149…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00093070 CAS Number: 1878-91-7 EINECS: 217-530-5 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...