2-Chloro-7H-purin-6-amine 97%
|
Warning
|
Synonyms:
6-Amino-2-chloropurine
, 2-Chloroadenine
H60869-ME
H60869-ME
AAH60869-MEEA
86.64
USD
CAAAH60869-ME
AAH60869-ME
2-Chloro-7H-purin-6-amine 97%
2-Chloro-7H-purin-6-amine
Formula:
C5H4ClN5 MW: 169.57 g/mol Melting Pt: 384…389 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037925 CAS Number: 1839-18-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...