(S)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl ≥97%

Warning

Synonyms: (S)-Xylyl-Garphos(S)-(4,4',6,6'-Tetramethoxybiphenyl-2,2'-diyl)bis[bis(3,5-dimethylphenyl)phosphine]

H60865-03 H60865-03 H60865-MD H60865-MD
AAH60865-03EA 501.96 USD
CAAAH60865-MD AAH60865-03 CAAAH60865-03 AAH60865-MD
(S)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl ≥97%
(S)-2,2'-Bis[bis(3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl
Formula: C₄₈H₅₂O₄P₂
MW: 754.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19443624
CAS Number: 1365531-90-3

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR