4'-(5-Chloro-2-methoxyphenylsulfamoyl)acetanilide 97%

Warning

H60820-03 H60820-MD H60820-03 H60820-MD
AAH60820-MDEA 69.2 USD
CAAAH60820-03 AAH60820-MD AAH60820-03 CAAAH60820-MD
4'-(5-Chloro-2-methoxyphenylsulfamoyl)acetanilide 97%
4'-(5-Chloro-2-methoxyphenylsulphamoyl)acetanilide
Formula: C₁₅H₁₅ClN₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00026154
CAS Number: 196869-06-4

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR