2,2,3,3-Tetramethylcyclopropanecarboxylic acid 97%
|
Warning
|
H60809-06
H60809-03
H60809-06
H60809-03
AAH60809-06EA
154.5
USD
CAAAH60809-06
CAAAH60809-03
AAH60809-06
AAH60809-03
2,2,3,3-Tetramethylcyclopropanecarboxylic acid 97%
2,2,3,3-Tetramethylcyclopropanecarboxylic acid
Formula:
C₈H₁₄O₂ MW: 142.20 g/mol Melting Pt: 120…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011538 CAS Number: 15641-58-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...