(2S,5S)-1-(2-[(2S,5S)-2,5-Diethyl-1-phospholanyl]phenyl)-2,5-diethylphospholane-1-oxide ≥97%

Synonyms: [1-(2S,5S)-2,5-Diethylphospholanyl]-[2-(2S,5S)-2,5-diethylphospholanyl-1-oxide]benzene

H60788-03 H60788-03 H60788-MD H60788-MD
AAH60788-03EA 397.38 USD
AAH60788-03 CAAAH60788-MD AAH60788-MD CAAAH60788-03
(2S,5S)-1-(2-[(2S,5S)-2,5-Diethyl-1-phospholanyl]phenyl)-2,5-diethylphospholane-1-oxide ≥97%
(2S,5S)-1-(2-[(2S,5S)-2,5-Diethyl-1-phospholanyl]phenyl)-2,5-diethylphospholane-1-oxide

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR