4-Methylhippuric acid 98%

Warning

Synonyms: 2-(4-Methylbenzamido)acetic acid

H60754-03 H60754-03
AAH60754-03EA 37.86 USD
AAH60754-03 CAAAH60754-03
4-Methylhippuric acid 98%
4-Methylhippuric acid
Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00020449
EINECS: 248-231-8

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR