2-Bromophenethylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(2-Bromophenyl)ethylamine
H60739-04
H60739-09
H60739-09
H60739-04
AAH60739-04EA
74.09
USD
AAH60739-04
CAAAH60739-04
CAAAH60739-09
AAH60739-09
2-Bromophenethylamine 97%
2-Bromophenethylamine
Formula:
C₈H₁₀BrN Boiling Pt: 253 °C Density: 1.396 Storage Temperature: Ambient |
MDL Number:
MFCD01529874 UN: 2735 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...