3,4,6-Tri-O-benzyl-2-deoxy-D-glucono-1,5-lactone 97%

Supplier: Thermo Scientific Chemicals

H60716-MD H60716-03 H60716-MD H60716-03
AAH60716-03EA 167.99 USD
CAAAH60716-MD AAH60716-03 CAAAH60716-03 AAH60716-MD
3,4,6-Tri-O-benzyl-2-deoxy-D-glucono-1,5-lactone 97%
3,4,6-Tri-O-benzyl-2-deoxy-D-glucono-1,5-lactone
Formula: C₂₇H₂₈O₅
MW: 432.516 g/mol
Melting Pt: 83…88 °C
Storage Temperature: Ambient
MDL Number: MFCD22989012
CAS Number: 72605-77-7

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR