5-Chloro-2-methylbenzoic acid 98%

Warning

Synonyms: 5-Chloro-2-methylbenzoic acid

H60649-03 H60649-03 H60649-06 H60649-06
AAH60649-06EA 59.99 USD
AAH60649-06 AAH60649-03 CAAAH60649-03 CAAAH60649-06
5-Chloro-2-methylbenzoic acid 98%
5-Chloro-2-methylbenzoic acid
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Melting Pt: 168…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00045842
CAS Number: 7499-06-1

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR