2-Chloro-4-methylpyridine 98%

Warning

Synonyms: 2-Chloro-4-picoline

H60607-AC H60607-AA H60607-AA H60607-AC
AAH60607-ACEA 293.83 USD
AAH60607-AC AAH60607-AA CAAAH60607-AC CAAAH60607-AA
2-Chloro-4-methylpyridine 98%
2-Chloro-4-methylpyridine
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 194…195 °C
Melting Pt: 115 °C
Density: 1.142
Storage Temperature: Ambient
MDL Number: MFCD00023418
CAS Number: 3678-62-4
EINECS: 222-951-2

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR