(S)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl ≥97%
|
Warning
|
Synonyms:
(S)-BTFM-Garphos
H60604-03
H60604-MD
H60604-03
H60604-MD
AAH60604-03EA
541.68
USD
CAAAH60604-03
CAAAH60604-MD
AAH60604-03
AAH60604-MD
(S)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl ≥97%
(S)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl
Formula:
C₄₈H₂₈F₂₄O₄P₂ MW: 1186.64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19443625 CAS Number: 1365531-85-6 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...