5-Bromo-2-fluorobenzoic acid 97%

Warning

H60595-06 H60595-03 H60595-03 H60595-06
AAH60595-06EA 115.85 USD
CAAAH60595-03 CAAAH60595-06 AAH60595-06 AAH60595-03
5-Bromo-2-fluorobenzoic acid 97%
5-Bromo-2-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00143423
CAS Number: 146328-85-0
ADR: 6.1,III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR