3,4-Di-O-acetyl-D-glucal 97%

Supplier: Thermo Scientific Chemicals

H60585-MD H60585-03 H60585-MD H60585-03
AAH60585-MDEA 62.48 USD
CAAAH60585-MD CAAAH60585-03 AAH60585-MD AAH60585-03
3,4-Di-O-acetyl-D-glucal 97%
3,4-Di-O-acetyl-D-glucal
Formula: C₁₀H₁₄O₆
MW: 230.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22989007
CAS Number: 82295-98-5

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR