N,N-Diethyl-4-nitrobenzamide 97%

Warning

H60565-MD H60565-03 H60565-03 H60565-MD
AAH60565-MDEA 152.07 USD
AAH60565-MD CAAAH60565-03 CAAAH60565-MD AAH60565-03
N,N-Diethyl-4-nitrobenzamide 97%
N,N-Diethyl-4-nitrobenzamide
Formula: C₁₁H₁₄N₂O₃
MW: 222.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00454141
CAS Number: 5323-47-7

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR