2,3,6-Trimethylphenol 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Hydroxypseudocumene
H60489-14
H60489-22
H60489-22
H60489-14
AAH60489-14EA
28.79
USD
AAH60489-14
AAH60489-22
CAAAH60489-14
CAAAH60489-22
2,3,6-Trimethylphenol 95%
2,3,6-Trimethylphenol
Formula:
C₉H₁₂O MW: 136.19 g/mol Boiling Pt: 215 °C Melting Pt: 59…62 °C Density: 0.940 Flash Pt: 100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002229 CAS Number: 2416-94-6 EINECS: 219-330-3 UN: 2430 ADR: "8",III |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...