(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(Rp)-1-[(1S)-(1-Aminoethyl)]-2-(diphenylphosphino)ferrocene
H60488-MD
H60488-03
H60488-MD
H60488-03
AAH60488-03EA
596.07
USD
CAAAH60488-03
AAH60488-03
CAAAH60488-MD
AAH60488-MD
(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine ≥97%
(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine
Formula:
C₂₄H₂₄FeNP MW: 413.27 g/mol Melting Pt: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD11045439 CAS Number: 607389-84-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...