1,1'-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ferrocene ≥97%
Synonyms:
(R,R)-Isopropyl-Ferrocelane
H60460-MD
H60460-03
H60460-MD
H60460-03
AAH60460-MDEA
126.54
USD
AAH60460-MD
CAAAH60460-03
AAH60460-03
CAAAH60460-MD
1,1'-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ferrocene ≥97%
1,1'-Bis[(2R,5R)-2,5-diisopropyl-1-phospholanyl]ferrocene
Formula:
C₃₀H₄₈FeP₂ MW: 518,43 g/mol Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD09750465 CAS Number: 849950-54-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...