DL-Leucic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Hydroxy-4-methylvaleric Acid

H60459-03 H60459-03
AAH60459-03EA 68.99 USD
CAAAH60459-03 AAH60459-03
DL-Leucic acid 98%
DL-Leucic acid
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00004246
CAS Number: 498-36-2
EINECS: 207-860-8

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR