3-Bromo-N-methylbenzenesulfonamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H60436-03 H60436-03 H60436-MD H60436-MD
AAH60436-MDEA 80.89 USD
CAAAH60436-03 AAH60436-MD CAAAH60436-MD AAH60436-03
3-Bromo-N-methylbenzenesulfonamide 97%
3-Bromo-N-methylbenzenesulphonamide
Formula: C₇H₈BrNO₂S
MW: 250.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363818
CAS Number: 153435-79-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR