2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals

H60431-MD H60431-03 H60431-03 H60431-MD
AAH60431-03EA 360.99 USD
AAH60431-03 AAH60431-MD CAAAH60431-03 CAAAH60431-MD
2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester 96%
2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester
Formula: C₁₂H₁₂BF₅O₂
MW: 294.028 g/mol
Boiling Pt: 66 °C/0.4 mm
Melting Pt: 35…36 °C
Storage Temperature: Ambient
MDL Number: MFCD12405352
CAS Number: 325142-81-2

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR